Learn More
Reichert™ NIST Certified Calibration Oil
Check the accuracy of an instrument using calibration oils. Reichert™ NIST Certified Calibration Oil are formulated laser liquid calibration oils that are supplied with an NIST certification of traceability.
Brand:  Reichert™ 13K41390
 View more versions of this product
Product Code. 16317651
  • 484.00 € / Each
Explore more special offers
Description
Description
Formulated, measured, and certified to five significant places of refractive index (0.00001).
Specifications
Specifications
NIST Certified Calibration Oil
1.35999
NIST certification of traceability
18.04 Brix
Refractometer