Learn More
Nitazoxanide-d4, MedChemExpress
MedChemExpress Nitazoxanide D4 (NTZ D4) is the deuterium labeled Nitazoxanide, which is an antiprotozoal agent.
Brand: Medchem Express HY-B0217S
This item is not returnable.
View return policy
Description
Nitazoxanide D4 (NTZ D4) is the deuterium labeled Nitazoxanide, which is an antiprotozoal agent.Specifications
Nitazoxanide-d4 | |
10 mg | |
C12H5D4N3O5S | |
NTZ-d4 NSC-697855-d4 | |
O=C(NC1=NC=C([N+]([O-])=O)S1)C2=C(OC(C)=O)C([2H])=C([2H])C([2H])=C2[2H] | |
311.31 | |
Research |
1246819-17-9 | |
Research | |
Solid | |
Off-White | |
311.31 | |
98.0% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Nitazoxanide-d4, MedChemExpress