missing translation for 'onlineSavingsMsg'
Learn More

Nitenpyram, Thermo Scientific Chemicals

Product Code. 16594394
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150824-47-8
Molecular Formula C11H15ClN4O2
Molecular Weight (g/mol) 270.72
InChI Key CFRPSFYHXJZSBI-DHZHZOJOSA-N
IUPAC Name [(1E)-1-{[(6-chloropyridin-3-yl)methyl](ethyl)amino}-2-nitroethenyl](methyl)amine
SMILES CCN(CC1=CC=C(Cl)N=C1)C(\NC)=C\[N+]([O-])=O

Specifications

Quantity 250 mg
Flash Point >70°C
Melting Point 70.0°C to 84.0°C
Solubility Soluble in water; Insoluble in methanol and dimethyl sulfoxide
Formula Weight 270.72
Chemical Name or Material Nitenpyram

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitenpyram, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.