missing translation for 'onlineSavingsMsg'
Learn More

Nitinol foil, 0.05mm (0.002in) thick, shape memory, flat annealed, pickled surface, Thermo Scientific Chemicals

102.51 € - 1037.16 €

Specifications

Chemical Name or Material Nitinol foil
Name Note 0.05mm (0.002 in.) thick, shape memory, flat annealed, pickled surface
MDL Number MFCD02091734
Solubility Information Insoluble in water.
Odor Odorless
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424015
View Documents
Thermo Scientific Alfa Aesar
045663.FF
25 x 25 mm
102.51 €
Each
Please to purchase this item. Need a web account? Register with us today!
15434015
View Documents
Thermo Scientific Alfa Aesar
045663.FI
50 x 50 mm
291.45 €
Each
Please to purchase this item. Need a web account? Register with us today!
15444015
View Documents
Thermo Scientific Alfa Aesar
045663.GE
75 x 100 mm
1037.16 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nitinol foil is used in cell phone antennas, children's toys, experimental materials, eye glasses components and medical equipment. It is also used in heat engines, resilient glass frames, mechanical watch springs, temperature control system, couplings, golf club insert, biomedical and medical uses, cell-phone technology: retractable antenna, microphone boom. It is highly biocompatible and has properties suitable for use in orthopedic implants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nitinol foil
MFCD02091734
Odorless
0.05mm (0.002 in.) thick, shape memory, flat annealed, pickled surface
Insoluble in water.
≈0.25g/25x25mm
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.