missing translation for 'onlineSavingsMsg'
Learn More
Nitrate-free Water, Fisher Chemical™ Product Code.: 12980564

Nitrate-free Water, Fisher Chemical™

Product Code. 12980564
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy

Product Code. 12980564

Brand: Fisher Chemical RP 230 1000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Quantity 1000 mL
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1104134 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberRP 230 1000
Lot Number1101073 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog NumberRP 230 1000
Lot Number1096007 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberRP 230 1000
Lot Number1090794 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberRP 230 1000
Lot Number1092916 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberRP 230 1000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitrate-free Water, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.