missing translation for 'onlineSavingsMsg'
Learn More

Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11373647
Click to view available options
Quantity:
100 mL

Nitrate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00144920

Specifications

Quantity 100 mL
Grade Specpure™
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Nitrate Ion chromatography standard solution, NO3- 1000μg/mL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.