missing translation for 'onlineSavingsMsg'
Learn More

Nitrate ISE Standard Solution, Mettler Toledo™

Product Code. 11917531
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Description ISE standard NO3 1000 mg/L, 500mL
Packaging 500 mL
Quantity 500 mL
Color Colorless
Physical Form Liquid
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'