Learn More
WTW™ Nitrate Test Kits
Brand:  WTW™ 250411
Product Code. 11761093
  • 193.00 € / Pack of 25
Explore more special offers
Specifications
Phenol derivative
PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
45
Photometric
For waste water, drinking water, surface water
Certificates
25 Tests
CombiCheck Nitrate (NO3) cell test kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • nitrate cell test in seawater
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, phosphoric acid