Learn More
Nitric acid, 2.0N Standardized Solution, Thermo Scientific Chemicals
Description
Nitric acid, 2.0N Standardized Solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 1.06 g/mL |
Quantity | 1 L |
Physical Form | Liquid |
Solubility Information | Miscible with water and ether. |
Formula Weight | 63.01 |
Chemical Name or Material | Nitric Acid, 2N Standardized Solution |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
62400751 | Certificate of Analysis | 17/02/2025 | 044528.AP,044528.K2 |
62400443 | Certificate of Analysis | 17/02/2025 | 044528.AP,044528.K2 |
M16L528 | Certificate of Analysis | 27/01/2025 | 044528.AP,044528.K2 |
Y29K505 | Certificate of Analysis | 26/12/2024 | 044528.AP,044528.K2 |
X02K510 | Certificate of Analysis | 13/11/2024 | 044528.AP,044528.K2 |
Safety and Handling

- Nitric acid, 2.0N Standardized Solution
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Nitric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.