Learn More
Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
Description
In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuelsMain commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless to Yellow |
Boiling Point | 121°C |
Density | 1.41 g/cm3 at 20°C |
Odor | Pungent |
Vapor Pressure | 17mm Hg at 20°C |
Assay Percent Range | 99.999% (metals basis) |
Quantity | 500 mL |
Physical Form | Liquid |
UN Number | 2031 |
Solubility Information | Very soluble in water and ether. Decomposes in alcohol.Miscible with water and ether. |
Show More |
Safety and Handling



- Nitric acid, 65-70%
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 3
- Skin corrosion/irritation Category 1 A
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Nitric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.