missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals

Product Code. p-3598428 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic bottle
Unit Size:
1 L
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10485993

Brand: Thermo Scientific Acros 124660011

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2, 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Melting Point -42.0°C
Boiling Point 122.0°C
CAS Min % 64.0, 34.0
CAS Max % 66.0, 36.0
Density 1.4000g/mL
Packaging Plastic bottle
Specific Gravity 1.4
Quantity 1 L
Linear Formula HNO3
Physical Form Liquid
Fieser 01,733; 03,212; 04,356; 05,474
Merck Index 15, 6663
Solubility Information Solubility in water: >100 g/100mL (20°C).
Formula Weight 63.01
Percent Purity ≥65%
Grade Analytical
Chemical Name or Material Nitric acid
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.