missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Nitrile Gloves

Product Code. 12635283
Click to view available options
Size:
7
8
9
Color:
Green
Model:
948
953
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12635283

Brand: Honeywell Safety Products™ 209530107

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Chemically resistant nitrile gloves with a patterned finish to provide grip. Honeywell Safety Products™ Powercoat™ Nitraf Gloves have a flocked cotton interior and a protective cuff.

  • Nitrile composition for mechanical and chemical resistance to a large range of products including oils, greases, hydrocarbons, detergents and many solvents.
  • Avoid contact with solvents containing ketones.
  • Suitable for food preparation that includes greasy products, production of paints, varnishes.
  • Flocked-cotton inside for comfort and easy donning and doffing.
  • Raised pattern finish for good grip in greasy and damp environments.
  • Great elasticity and dexterity.
  • Safety Category III glove.
  • Cuff helps to protect a large part of the forearm.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY III Irreversible risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical risks, EN 374-2003 Chemical risk, EC Category PPE 3, EC Certificate Number 0075/797/162/02/07/0042, EC Attestation Number 0075/797/162/02/07/0042
Classification Dexterity: 5
Color Green
Cut Resistance Rating 1
Material Nitrile, Cotton
Product Type Gloves
Size 7
Thickness (Metric) 0.4 mm
For Use With (Application) Agriculture, Automotive, Aviation, Building, Construction, Catering, Chemical, Food, Industrial Cleaning, Iron, Steel, Maintenance, Offshore, Petrochemical, Printing, Service, Ship Building, Transportation, Utility and Water Treatment industries.
Liner Material Cotton Flock
Lining Lined
Model 953
Puncture Rating 1
Quantity 1 Pair/PK
Resistant To Chemicals, Oils
Tear Rating 0
Length (Metric) 33 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.