missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Nitritex™ Omega Pro-tech™ Blue Disposable Powder-free Laboratory Gloves

Blue disposable powder free gloves resist cuts and snags and are available in a variety of sizes.

22.05 €

Specifications

Certifications/Compliance ISO 2859
Classification Category 3 - Complex Design Personal Protective Equipment
Color Blue
Disposable Single-use
Latex Free Yes
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12910845
View Documents
Nitritex™
OTEBL2103-M
Medium
22.05 €
Pack of 100
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
12930845
View Documents
Nitritex™
OTEBL2105-XL
X-Large
22.05 €
Pack of 100
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
12920845
View Documents
Nitritex™
OTEBL2104-L
Large N/A N/A N/A
12900845
View Documents
Nitritex™
OTEBL2102-S
Small
22.05 €
Pack of 100
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
Description

Description

  • Blue
  • Beaded cuff
  • Non-sterile
  • Nitrile contains no proteins
  • Various sizes
Specifications

Specifications

ISO 2859
Blue
Yes
Nitrile Examination Glove
Yes
Category 3 - Complex Design Personal Protective Equipment
Single-use
Nitrile
Beaded/Rolled
24 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitritex™ Nitritex™ Omega Pro-tech™ Blue Disposable Powder-free Laboratory Gloves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.