missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrilotriacetic acid, Honeywell Fluka™
Click to view available options
Quantity:
50 g
6 x 50 g
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Flash Point | 100°C (212°F) |
Linear Formula | N(CH2COOH)3 |
Quantity | 50 g |
UN Number | NONH for all modes of transport |
Beilstein | 1710776 |
Formula Weight | 191.14g/mol |
Percent Purity | ≥99% |
Chemical Name or Material | Nitrilotriacetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'