Learn More
Nitrite, Quant™ Test Strips, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 023283.QA
Product Code. 15411137
  • 162.81 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Measures 1 to 50ppm NO2- in aqueous solutio
Nitrite test strip