missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Nitrite Test Kits
Obtain precision measurements for waste water, ground and surface water, drinking, sea water, food and soils after corresponding sample preparation with the Nitrite (NO2) cell test kits.
Brand: WTW™ 252074
This item is not returnable.
View return policy
Description
- Nitrite (NO<sub>2</sub>) cell test kit
- This test is highly accurate and easy to use.
- For use with PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
- For waste water, ground and surface water, drinking water, sea water, food and soils after corresponding sample preparation
- Griess Reaction Method
- Photometric Test Method
Specifications
Griess Reaction | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
25 | |
Photometric |
For waste water, ground and surface water, drinking water, sea water, food and soils after corresponding sample preparation | |
Certificates | |
25 Tests | |
Nitrite (NO2) cell test kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Nitrite Cell Test N5/25 Method: photometric 0.010 - 0.700 mg/l NO
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sulfanilic acid, N-(1-Naphthyl)ethylenediamine dihydrochloride