missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Nitrocellulose Blotting Membrane

Ideal for Western blotting, DNA blotting as well as dot or slot blots. 0.45 μm nitrocellulose blotting membranes are optimised for all protein blotting systems, such as electrotransfer, semi-dry or simple capillary blotting.

Brand:  Sartorius™ 11306------41BL

345.80 EUR valid until 2025-12-31
Use promo code "24363" to get your promotional price.



Product Code. 10732793

  • 364.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High membrane surface area for high binding capacity and no sample loss
  • Exceptionally low background allowing longer exposure time and better results
  • High membrane stability for easy handling
TRUSTED_SUSTAINABILITY
Specifications

Specifications

30 mm
Optimally suited for dot or slot blot
>200 μg/cm2; IgG
0.45 μm
300 mm
Western Blotting
1 Each
30 x 300 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CN 0.45 micron, Roll 300mm x 3m,Blotting

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P230-Keep wetted with .
  • P243-Take precautionary measures against static discharge.
  • P234-Keep only in original container.
  • P233-Keep container tightly closed.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P372-Explosion risk in case of fire.
  • P375-Fight fire remotely due to the risk of explosion.
  • P370+P380-In case of fire: Evacuate area.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Nitrogen in nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.