missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Nitrocellulose Membrane for Blotting
Sartorius Stedim Biotech™ Nitrocellulose Membrane for Blotting combines the advantages of high protein binding capacity with low background and high membrane stability.
Brand: Sartorius Stedim Biotech™ 11327------41BL
258.50 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- High protein binding capacity.
- Low background and high membrane stability.
- Easy handling.
- Large surface area.
- Ideal for western blotting, DNA blotting, dot blots, slot blots.
- Optimized for all protein blotting systems such as electrotransfer, semi-dry, or simple capillary blotting.
Specifications
Membrane for blotting | |
Nitrocellulose | |
130 μm |
Cellulose Nitrate | |
0.22 μm | |
≤1s |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Nitrocellulose Blotting Membrane 0.22um, Roll 300m x 3m
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose