missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nitrocellulose Membranes, 0.2 μm

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods).

172.00 € - 424.00 €

Specifications

Format Sheet
Material Nitrocellulose
Pore Size 0.2 μm
Shipping Condition Room Temperature
View More Specs

Products 3
Product Code Brand Dimensions (LxW) Format Pore Size Sufficient For Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Format Pore Size Sufficient For Price Quantity & Availability  
10148113
View Documents
Thermo Scientific™
88013
10.5 x 7.9 cm Sheet 0.2 μm 15 Mini-gel Blots
195.00 €
Pack of 15
Please to purchase this item. Need a web account? Register with us today!
10096363
View Documents
Thermo Scientific™
88024
8 x 8 cm Sheet 0.2 μm 15 Mini-gel Blots
172.00 €
Pack of 15
Please to purchase this item. Need a web account? Register with us today!
10561674
View Documents
Thermo Scientific™
77012
12 x 8 cm Sheet 0.2 μm 25 Mini-gel Blots
424.00 €
0.25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods). The 0.2-μm pore size is well-suited for western blotting, and this smaller pore size is ideal for the transfer of low molecular weight proteins or peptides and nucleic acids (<300 bp).

Features include:
100% pure nitrocellulose membrane
• Convenient—save time with pre-cut sheets
• Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• High-binding affinity—provides excellent protein binding, blocks easily, and does not require pre-wetting with alcohol
• Low background—produce high-quality data with low background signal in both chemiluminescent and fluorescent western blotting

Specifications

Specifications

Sheet
Nitrocellulose
0.2 μm
Room Temperature
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nitrocellulose Membranes, 0.2 μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.