Learn More
Thermo Scientific™ Nitrocellulose Membranes, 0.45 μm
Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods).
172.00 € - 540.00 €
Specifications
Material | Nitrocellulose |
---|---|
Pore Size | 0.45 μm |
Shipping Condition | Room Temperature |
Product Code | Brand | Dimensions (LxW) | Format | Pore Size | Sufficient For | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Format | Pore Size | Sufficient For | Price | Quantity & Availability | ||
10270584
|
Thermo Scientific™
88025 |
8 x 8 cm | Sheet | 0.45 μm | 15 Mini-gel Blots |
172.00 €
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10551584
|
Thermo Scientific™
88014 |
10.5 x 7.9 cm | Sheet | 0.45 μm | 15 Mini-gel Blots |
193.00 €
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10168543
|
Thermo Scientific™
77010 |
12 x 8 cm | Sheet | 0.45 μm | 25 Mini-gel Blots |
412.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10773485
|
Thermo Scientific™
88018 |
3.5 m x 30 cm | Roll | 0.45 μm | ≥120 Mini-gel Blots (When cut to 8 x 10 cm) |
540.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods). The 0.45-μm pore size is well-suited for a wide range of western blotting conditions and ideal for the transfer of most proteins (>20 kDa) and nucleic acids (>300 bp).
Features include:
• 100% pure nitrocellulose membrane
• Convenient—save time with pre-cut sheets or choose the roll format for the flexibility to cut to the ideal size
• Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• High-binding affinity—provides excellent protein binding, blocks easily, and does not require pre-wetting with alcohol
• Low background: produce high-quality data with low background signal in both chemiluminescent and fluorescent western blotting
Specifications
Nitrocellulose | |
0.45 μm | |
Room Temperature |
Safety and Handling
- NITROCELLULOSE MEMBRANE, 0.45U
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
- MIXTURE LIST-Contains : Nitrocellulose
Your input is important to us. Please complete this form to provide feedback related to the content on this product.