missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Nitrocellulose stripping buffer (10X)
59.09 € - 88.40 €
Specifications
Product Type | Western Blot Stripping Buffer |
---|---|
Form | Liquid |
Concentration | 10X |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15414909
|
Thermo Scientific Chemicals
J62541.AK |
250 mL |
59.09 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15424909
|
Thermo Scientific Chemicals
J62541.AP |
500 mL |
88.40 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ideal for removal of secondary antibody after detection of highly abundant targets. Nitrocellulose Stripping Buffer may perform well with nitrocellulose membranes or antibody-dye conjugates. It is also employed for nucleic acid and protein related blotting works.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Western Blot Stripping Buffer | |
Liquid | |
10X |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only