missing translation for 'onlineSavingsMsg'
Learn More

Aalborg™ Nitrogen Direct Flow Tube 150mm Reference Scale

Aalborg™ Nitrogen Direct Flow Tube 150mm Reference Scale completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.

Brand:  Aalborg™ FT-112-06-SA-VN

 View more versions of this product

Product Code. 11758288

  • 118.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications

Specifications

500 mL/min.
Nitrogen Direct Flow Tube
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.