missing translation for 'onlineSavingsMsg'
Learn More

Nitromathan in Dimethyl Sulfoxide, SPEX CertiPrep™

Product Code. 16369757
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Nitromethane75-52-5250μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 75-52-5
Packaging 2mL amber prescored glass ampule
Quantity 1 mL
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula CH3NO2
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
SMILES C[N+]([O-])=O
IUPAC Name nitromethane
Molecular Weight (g/mol) 61.04
PubChem CID 6375
ChEBI CHEBI:77701
Explosion Limit 1.8% (volume); 63% (volume)
Grade USP
Concentration or Composition (by Analyte or Components) 250μg/mL
Ignition Point 270°C
Vapor Pressure 2.5 hPA (20°C)
Analyte or Component Names Nitromethane
Density 1.1g/cm3
Solvent or Matrix Dimethyl Sulfoxide
Physical Form Fluid
Chemical Name or Material Nitromethane in Dimethyl Sulfoxide, USP 467 Standard
Show More Show Less
compliance-icons
Product Identifier
  • Stock VOA
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contains : dimethyl sulfoxide

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'