missing translation for 'onlineSavingsMsg'
Learn More

Nitronium hexafluorophosphate

Product Code. p-4824856
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11342248

Brand: Thermo Scientific Alfa Aesar 013151.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19200-21-6 | F6NO2P | 190.969 g/mol

Nitronium hexafluorophosphate is a nitrating reagent used in the nitration of alkanes such as methane, ethane, propane, n-butane, isobutane, neopentane and cyclohexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19200-21-6
Molecular Formula F6NO2P
Molecular Weight (g/mol) 190.969
MDL Number MFCD00031520
InChI Key DXWZKTLGZWTYPL-UHFFFAOYSA-N
Synonym nitronium hexafluorophosphate, nitronium ion hexafluorophosphate, dioxidonitrogen 1+ hexafluorophosphate
PubChem CID 45933661
IUPAC Name nitronium;hexafluorophosphate
SMILES [N+](=O)=O.F[P-](F)(F)(F)(F)F

Specifications

Color White
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in dichloromethane and nitroethane.
Formula Weight 190.97
Sensitivity Moisture sensitive
Appearance Off-white Cristals
Chemical Name or Material Nitronium hexafluorophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.