missing translation for 'onlineSavingsMsg'
Learn More

Nitronium tetrafluoroborate, 0.5M solution in sulfolane, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 175132000

603.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Product Code. 10732221

  • 606.02 € / 200mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitronium tetrafluoroborate
Colorless through Yellow
5.76, 94.97
200 mL
NO2BF4
01,742; 04,358; 05,476; 08,361; 09,324; 06,520; 07,324; 09,414; 12,435; 13,274; 14,282; 17,314
Solubility in water: reacts
[O-][NH+]=O.F[B-](F)(F)F
132.81
132.81
1.268
13826-86-3, 126-33-0
5.03, 94.24
Solution
BF4NO2
MFCD00011432
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
XYRQURYIJKJVSF-UHFFFAOYSA-N
nitronium;tetrafluoroborate
11073463
Glass bottle
1.2680g/mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Nitronium tetrafluoroborate, 0.5 M solution in sulfolane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 1B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only