missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitronium tetrafluoroborate, 97%, Thermo Scientific Chemicals
62.60 € - 553.00 €
Chemical Identifiers
CAS | 13826-86-3 |
---|---|
Molecular Formula | BF4NO2 |
Molecular Weight (g/mol) | 132.81 |
MDL Number | MFCD00011432 |
InChI Key | XYRQURYIJKJVSF-UHFFFAOYSA-N |
Synonym | nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane |
PubChem CID | 11073463 |
IUPAC Name | nitronium;tetrafluoroborate |
SMILES | [O-][NH+]=O.F[B-](F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10516603
|
Thermo Scientific Acros
174380050 |
5 g | Plastic bottle |
62.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10082370
|
Thermo Scientific Acros
174380250 |
25 g | Plastic bottle |
198.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904131
|
Thermo Scientific Acros
174381000 |
100 g | Plastic bottle |
553.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13826-86-3 | |
132.81 | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
11073463 | |
[O-][NH+]=O.F[B-](F)(F)F |
BF4NO2 | |
MFCD00011432 | |
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
nitronium;tetrafluoroborate |
Specifications
200.0°C | |
Colorless through White | |
96% min | |
NO2BF4 | |
01,742; 04,358; 05,476; 08,361; 09,324; 01,747; 04,360; 07,253; 08,364; 14,215 | |
Solubility in water: may decompose | |
[O-][NH+]=O.F[B-](F)(F)F | |
132.81 | |
132.81 | |
Nitronium tetrafluoroborate |
13826-86-3 | |
Adhering Crystals | |
BF4NO2 | |
MFCD00011432 | |
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
nitronium;tetrafluoroborate | |
11073463 | |
97% |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-533-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Nitronium tetrafluoroborate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only