missing translation for 'onlineSavingsMsg'
Learn More
Nitrosonium hexafluorophosphate, 95%, Thermo Scientific Chemicals Product Code.: 10655073

Nitrosonium hexafluorophosphate, 95%, Thermo Scientific Chemicals

Product Code. 10655073
5 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10655073

Brand: Thermo Scientific Acros 383730050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16921-91-8
Molecular Formula F6NOP
Molecular Weight (g/mol) 174.97
MDL Number MFCD00040324
InChI Key SAKPNYRUBHJGAR-UHFFFAOYSA-N
Synonym nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium
PubChem CID 10910083
IUPAC Name azanylidyneoxidanium;hexafluorophosphate
SMILES N#[O+].F[P-](F)(F)(F)(F)F

Specifications

Color Colorless or White
CAS Min % 94.0
CAS Max % 100.0
Physical Form Crystals
Quantity 5 g
Assay Percent Range 94% min
Solubility Information Solubility in water: decomposes
Formula Weight 174.97
Percent Purity 95%
Packaging Plastic bottle
Chemical Name or Material Nitrosonium hexafluorophosphate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453449 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number383730050,383730250
Lot NumberA0456952 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number383730050,383730250
Lot NumberA0440475 Certificate TypeCertificate of Analysis Date02/10/2023 Catalog Number383730050,383730250
Lot NumberA0453449 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number383730050,383730250
Lot NumberA0432081 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog Number383730050,383730250
compliance-icons
Product Identifier
  • Nitrosonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitrosonium hexafluorophosphate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.