missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium tetrafluoroborate, 97%

Product Code. 10135442 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10135442

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14635-75-7 | BF4HNO2 | 133.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14635-75-7
Molecular Formula BF4HNO2
Molecular Weight (g/mol) 133.82
MDL Number MFCD00011433
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate
PubChem CID 11137142
IUPAC Name azanylidyneoxidanium;tetrafluoroborate
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Melting Point 250.0°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 96% min. (ex F)
Linear Formula NOBF4
Merck Index 15, 6736
Formula Weight 116.81
Percent Purity 97%
Packaging Plastic bottle
Specific Gravity 2.185
Density 2.1850g/mL
Chemical Name or Material Nitrosonium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.