Learn More
NLN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA588992
Product Code. 16517051
  • 495.00 € / 100µL
Explore more special offers
Description
Description
Positive Samples: U-87MG, A-549, Rat liver; Cellular Location: Cytoplasm, Mitochondrion intermembrane space Immunogen sequence: DGQYYGYLWS EVFSMDMFYS CFKKEGIMNP EVGMKYRNLI LKPGGSLDGM DMLHNFLKRE PNQKAFLMSR GLHAP
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NLN
Polyclonal
Unconjugated
Nln
4930472G13Rik, AGTBP, angiotensin binding protein, angiotensin-binding protein, C79345, endopeptidase 24.16, EP24.16, KIAA1226, MEP, microsomal endopeptidase, Mitochondrial oligopeptidase M, MOP, neurolysin, neurolysin (metallopeptidase M3 family), neurolysin, mitochondrial, Neurotensin endopeptidase, NLN
Rabbit
Affinity Chromatography
RUO
117041, 57486
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1.91 mg/mL
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
P42676, Q9BYT8
Nln
Recombinant fusion protein containing a sequence corresponding to amino acids 630-704 of human NLN (NP_0657771)
100 μL
Primary
Human, Rat