Learn More
NLN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA598844
Product Code. 16591662
  • 444.00 € / 100µg
Explore more special offers
Description
Description
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NLN
Polyclonal
Unconjugated
Nln
4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN
Rabbit
Protein G
RUO
57486
-20°C or -80°C if preferred
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
3.22 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q9BYT8
Nln
Recombinant Human Neurolysin, mitochondrial protein (109-241AA)