missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM107, MedChemExpress
MedChemExpress NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.
95.85 € - 317.25 €
Specifications
Chemical Name or Material | NM107 |
---|---|
CAS | 20724-73-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C10H15N3O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16493988
|
Medchem Express
HY-10468 |
10 mg |
126.90 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16444996
|
Medchem Express
HY-10468 |
100 mg |
317.25 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16440347
|
Medchem Express
HY-10468 |
50 mg |
255.15 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16416707
|
Medchem Express
HY-10468 |
5 mg |
95.85 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.Specifications
NM107 | |
White | |
C10H15N3O5 | |
DMSO : 100 mg/mL (388.74 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (194.37 mM) | |
257.24 | |
98.9% | |
Solid |
20724-73-6 | |
Research | |
2'-C-Methylcytidine NM-107 | |
NC(C=CN1[C@@H]2O[C@H](CO)[C@@H](O)[C@@]2(C)O)=NC1=O | |
257.24 | |
Research | |
COVID-19-anti-virus |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only