missing translation for 'onlineSavingsMsg'
Learn More

NMUR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen NMUR1
Concentration 0.13 mg/mL
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883344
View Documents Promotion Details
Proteintech
14619-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16873344
View Documents Promotion Details
Proteintech
14619-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Receptor for the neuromedin-U and neuromedin-S neuropeptides.
Specifications

Specifications

NMUR1
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human
O55040, Q9HB89
10316, 14767
NMUR1 Fusion Protein Ag6235
Primary
-20°C
NMUR1
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
FM 3, FM3, G protein coupled receptor 66, GPC R, GPR66, neuromedin U receptor 1, NMUR1
NMUR1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications