missing translation for 'onlineSavingsMsg'
Learn More

Manutan No entry sign, self adhesive vinyl, 100mm x 300mm

Brand:  Manutan A346804

Encompass_Preferred

Product Code. 12805329

  • 4.63 € / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Available in aluminum or self-adhesive vinyl.
  • Self-adhesive vinyl
  • Dimensions (H x W): 100mm x 300mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers