missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ No-Stain™ Protein Labeling Reagent

Product Code. 16351358
Click to view available options
No. of Reactions:
10 reactions
40 reactions
Gel Compatibility:
Any Gel Type and Gel Chemistry
Unit Size:
10 reactions
40 reactions
This item is not returnable. View return policy

Product Code. 16351358

Brand: Invitrogen™ A44717

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A fast, easy to use, covalent protein labeling reagent, applied to a membrane after gel transfer that provides sensitive, linear detection of protein for total protein normalization of western blotting data. The Invitrogen No-Stain Reagent can also be used as a fast, sensitive gel stain for visualization of proteins in a gel.

The Invitrogen No-Stain Protein Labeling Reagent provides a flexible, accurate, rapid, and reliable method to visualize and normalize proteins in a gel or on a membrane (post-transfer). It forms covalent bonds to proteins in gels or on membranes within 10 minutes, does not require any de-staining steps, and can be instantly visualized using any commonly available imager. No-Stain reagent does not require any particular gels or other reagents and is compatible with gel stains and western workflows.

Instant visualization of proteins in gels

Coomassie and other gel staining and de-staining steps can be extremely time consuming and cumbersome. No-Stain Protein Labeling Reagent forms covalent bonds with the lysine amino acid side chains on all proteins in a gel within 10 minutes. Since lysine is one of the most abundant amino acids, No-Stain reagent enables detection of all proteins in a gel or on a membrane, and the strong signal emission from the covalently bonded reagent provides nanogram-level sensitivity.

  • Alternative to traditional gel staining reagents—provides more accurate normalization over a wide range of protein lysate concentrations (1–80 μg)
  • Sensitive—lower limit of detection of 20 ng per band
  • Specific—forms bonds only with the lysine side chains of proteins. Unbound reagent does not emit, thereby enabling a superior signal-to-noise ratio
  • Flexible—no need to change your gels to get stain-free convenience. No-Stain reagent provides stain-free convenience with any gel type (precast or pour-your-own gel)

Achieve the gold standard for quantitative western blotting

Protein normalization is a critical step in obtaining reliable and reproducible quantitative western blotting. Total protein normalization is considered the gold standard for quantitative western blotting. Many leading journals have developed guidelines for submitting western blotting research and select quotes from those guidelines are provided below.

  • 'For quantitative comparisons, appropriate reagents, controls and imaging methods with linear signal ranges should be used' – Nature
  • 'Record how data were obtained, whether signal intensity was linear with antigen loading, and how protein loading was normalized' – Journal of Biological Chemistry
  • 'Normalize signal intensity to total protein loading (assessed by staining membranes for total protein) whenever possible' – Journal of Biological Chemistry
  • 'House-keeping proteins should not be used for normalization without evidence that manipulations do not affect expression' – Journal of Biological Chemistry

An accurate loading control should display a linear relationship between signal intensity and sample load in all experimental conditions. The signal intensity obtained from labeling of total proteins on a membrane with No-Stain reagent ensures a linear relationship between signal intensity and sample load (see figure below) in all experimental conditions. Therefore, the use of No-Stain reagent in quantitative western blot applications enables the use of total protein as an ideal loading control.

  • Easy-to-use protocol—labeling of proteins within 10 minutes on either nitrocellulose or PVDF membranes
  • Rapid visualization using a wide-range of imagers with UV or fluorescence light sources
  • Accurate total protein normalization—the broad linear range for protein detection of 1–80 μg enables detection of No-Stain signal along with that of your target protein to achieve accurate total protein normalization
  • Sensitive and stable—nanogram level sensitivity with a stable signal that is compatible with downstream immunodetection steps
  • Superior analysis—housekeeping proteins are susceptible to signal saturation and other biological variations which are not observed when using No-Stain reagent for total protein normalization

Learn more about No-Stain Protein Labeling Reagent ›

TRUSTED_SUSTAINABILITY

Spécification

Green Features Less hazardous
Product Type Protein Labeling Reagent
Content And Storage • No-Stain activator, 200 μL, store at -5°C to -30°C
• No-Stain derivatizer, 200 μL, store at -5°C to -30°C
• No-Stain labeling buffer (20X), 2 x 5 mL, store at 15°C to 30°C
Description The No-Stain Protein Labeling Reagent forms covalent bonds with the lysine amino acid side chains on all proteins
For Use With (Equipment) Compatible with wide-range of imagers with UV or Fluorescence light sources, for example, the iBright imager
No. of Reactions 10 reactions
Run Time 10 minutes
Shipping Condition Dry Ice
Detection Method Fluorescence
Gel Compatibility Any Gel Type and Gel Chemistry
Membrane Compatibility Nitrocellulose, PVDF
Product Line Invitrogen
Quantity 1 kit
For Use With (Application) Compatible with all downstream immunodetection steps (blocking, antibody binding, chemiluminescent or fluorescence based detection
Afficher plus Afficher moins

For Research Use Only. Not for use in diagnostic procedures.

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis