missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOB1 Mouse anti-Human, Clone: 8C11A12, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | NOB1 |
---|---|
Clone | 8C11A12 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827575
|
Proteintech
66048-1-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16817575
|
Proteintech
66048-1-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5.8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.Specifications
NOB1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ART 4, ART4, MST158, MSTP158, NOB1, NOB1P, Protein ART 4, PSMD8BP1, RNA binding protein NOB1 | |
NOB1 | |
IgG1 | |
Protein G | |
Antibody |
8C11A12 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q9ULX3 | |
28987 | |
NOB1 Fusion Protein Ag17907 | |
Primary | |
-20°C | |
NOB1 |