missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nocodazole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 300.0°C to 305.0°C |
Color | Brown-Orange to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 10 mg |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in dmso,dmf,ethanol,chloroform,acetone |
Formula Weight | 301.32 |
Show More |
Safety and Handling


Product Identifier
- Nocodazole
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- H341-Suspected of causing genetic defects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nocodazole, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction