missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOLC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11815-1-AP-20UL
This item is not returnable.
View return policy
Description
NOPP140 (140 kDa nucleolar phosphoprotein) is implicated to play a role in ribosome assembly and the regulation of rRNA transcription. It has been observed to shuttle between the nucleus and cytoplasm and may also be important to nucleolar structural organization.Specifications
NOLC1 | |
Polyclonal | |
Unconjugated | |
NOLC1 | |
KIAA0035, NOLC1, NOPP130, NOPP140, NS5ATP13, Nucleolar 130 kDa protein, Nucleolar phosphoprotein p130 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64896, 70769, 9221 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
E9Q5C9, P41777, Q14978 | |
Nolc1 | |
NOLC1 Fusion Protein Ag2382 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction