missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA Classic

Product Code. 10223352
Click to view available options
Quantity:
100 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 10223352

Brand: QIAGEN WB120205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Classic with 4 sample areas per card.

  • QIAcard FTA Classic format with 4 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.

Specifications

Description Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Classic Cards.
Format Card
Sample Type Blood
For Use With (Equipment) Direct STR amplification
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples 4
Product Type FTA Classic Card
Quantity 100 Pk.
Sample Volume 125 mL
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Cellulose, sodium dodecyl sulphate, trometamol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.