Learn More
Non Ratio Turbidity Standard, Reagecon™
Use turbidity standards for precise measurements with nephelometeric instruments. Non Ratio Turbidity Standards, Reagecon™ are composed of suspended polymer microspheres to eliminate the problems with Formazin standards. They are NIST traceable, US EPA approved and certified accurate to ±1%.
78.70 € - 995.00 €
Specifications
Type | Non Ratio Turbidity Standard |
---|---|
For Use With (Equipment) | Non Ratio Instruments |
For Use With (Application) | Nephelometric turbidity measurement |
Accuracy | ±1% |
Certifications/Compliance | NIST, US EPA Approved |
Product Code | Brand | Volume (Metric) | Turbidity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Turbidity | Price | Quantity & Availability | ||||
11738505
|
Reagecon
CRS-10-100 |
100 mL | 10 NTU |
205.00 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11788495
|
Reagecon
CRS-0.0-500 |
500 mL | 0.0 NTU |
205.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11718515
|
Reagecon
CRS-100-100 |
100 mL | 100 NTU |
330.00 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11728515
|
Reagecon
CRS-100-500 |
500 mL | 100 NTU |
390.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11738515
|
Reagecon
CRS-200-100 |
100 mL | 200 NTU |
330.00 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11748515
|
Reagecon
CRS-200-500 |
500 mL | 200 NTU |
375.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11758505
|
Reagecon
CRS-20-100 |
100 mL | 20 NTU |
205.00 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11768505
|
Reagecon
CRS-20-500 |
500 mL | 20 NTU |
403.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11713836
|
Reagecon
CRS-4000-500 |
500 mL | 4000 NTU |
995.00 €
500mL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
||||
11703836
|
Reagecon
CRS-4000-100 |
100 mL | 4000 NTU |
397.00 €
100mL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
||||
11778505
|
Reagecon
CRS-40-100 |
100 mL | 40 NTU |
205.00 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11768515
|
Reagecon
CRS-500-500 |
500 mL | 500 NTU |
651.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11708515
|
Reagecon
CRS-50-500 |
500 mL | 50 NTU |
236.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11778515
|
Reagecon
CRS-800-100 |
100 mL | 800 NTU |
215.00 €
100mL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
||||
11788515
|
Reagecon
CRS-800-500 |
500 mL | 800 NTU |
375.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11728505
|
Reagecon
CRS-5-500 |
500 mL | 5 NTU |
205.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11748505
|
Reagecon
CRS-10-500 |
500 mL | 10 NTU |
403.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11708505
|
Reagecon
CRS-1-500 |
500 mL | 1 NTU |
205.00 €
500mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11778495
|
Reagecon
CRS-0.0-100 |
100 mL | 0.0 NTU |
78.70 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11718505
|
Reagecon
CRS-5-100 |
100 mL | 5 NTU |
88.00 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
11798495
|
Reagecon
CRS-1-100 |
100 mL | 1 NTU |
147.00 €
100mL |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
Descripción
These turbidity standards remove the handling, stability and accuracy problems associated with traditional Formazin turbidity standards. They are certified traceable to NIST Reference Material 1690, and are available in a range of concentrations from 0–4000 NTU.
- Non-toxic and non-carcinogenic, with no special handling or disposal requirements
- Well defined spherical shape of particles, 0.01–0.2μm in diameter
- Particles stay in suspension—they do not deteriorate or settle out
- A long stable shelf life of 2 years at all concentrations
- Convenient, ready-to-use solution—mixing is discouraged as it entrains air
Especificaciones
Non Ratio Turbidity Standard | |
Nephelometric turbidity measurement | |
NIST, US EPA Approved |
Non Ratio Instruments | |
±1% |
Seguridad y manipulación
missing translation for 'shelfLife' : 2 Years
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.