Learn More
Nonacarbonyldiiron, 99%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 15321-51-4 |
---|---|
Molecular Formula | C9H3Fe2O9 |
Molecular Weight (g/mol) | 366.804 |
MDL Number | MFCD00151465 |
InChI Key | CGJSNKHFIBINMR-UHFFFAOYSA-N |
Synonym | iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe |
PubChem CID | 71311424 |
IUPAC Name | carbon monoxide;iron;iron(3+);methanone |
SMILES | [CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3] |
Description
It is an important reagent in organometallic chemistry and of occasional use in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15321-51-4 | |
366.804 | |
CGJSNKHFIBINMR-UHFFFAOYSA-N | |
71311424 | |
[CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3] |
C9H3Fe2O9 | |
MFCD00151465 | |
iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe | |
carbon monoxide;iron;iron(3+);methanone |
Specifications
15321-51-4 | |
Orange | |
C9H3Fe2O9 | |
MFCD00151465 | |
Air Sensitive | |
Insoluble in water. | |
[CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3] | |
366.804 | |
363.79 | |
Crystalline |
∼100°C (decomposition) | |
2.85 g/mL | |
Fe2(CO)9 | |
UN3179 | |
iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe | |
CGJSNKHFIBINMR-UHFFFAOYSA-N | |
carbon monoxide;iron;iron(3+);methanone | |
71311424 | |
99% | |
Nonacarbonyldiiron |
Safety and Handling
GHS H Statement
H228-H301-H330
Flammable solid.
Toxic if swallowed.
Fatal if inhaled.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 239-359-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nonacarbonyldiiron
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
- Acute toxicity Category 3
- Acute toxicity Category 2
Hazard Statement
- H228-Flammable solid.
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only