missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonanoyl chloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | -61°C |
Density | 0.94 |
Boiling Point | 215°C |
Flash Point | 95°C (203°F) |
Refractive Index | 1.438 |
Quantity | 100 g |
UN Number | UN3265 |
Beilstein | 1362621 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'