missing translation for 'onlineSavingsMsg'
Learn More

Nonanoyl chloride, 96%, Thermo Scientific Chemicals

Product Code. 11493248
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 764-85-2
Molecular Formula C9H17ClO
Molecular Weight (g/mol) 176.684
MDL Number MFCD00000768
InChI Key NTQYXUJLILNTFH-UHFFFAOYSA-N
Synonym nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride
PubChem CID 69819
IUPAC Name nonanoyl chloride
SMILES CCCCCCCCC(=O)Cl

Specifications

Melting Point -61°C
Density 0.94
Boiling Point 215°C
Flash Point 95°C (203°F)
Refractive Index 1.438
Quantity 25 g
UN Number UN3265
Beilstein 1362621
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 176.69
Percent Purity 97%
Chemical Name or Material Nonanoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nonanoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Hazard Statement
  • H261-In contact with water releases flammable gases.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Nonanoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'