missing translation for 'onlineSavingsMsg'
Learn More

uvex NORA White 94766 Polymer Boot with PVC-Sole - Width 11

Long PVC safety boot made with steel toecap and midsole

Brand:  uvex 9476640

35.38 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 17217391

  • 39.80 € / 1 pair
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

NORA Safety Boot 9476.6 S5 SRC

  • Free of plasticizers included on SVHC Candidate List of the REACH regulation
  • Resistant to oil, grease, fuel, alkalis, acids and disinfectants
  • Anatomically shaped lasts, prevent fatigue
  • Long shaft can be shortened to meet individual requirements
    • Shaft height: 33 cm (for size 42 boot)
  • Anatomically shaped footbed
  • Toe and heel reinforcements
  • Ankle protection
  • Ribbed heel with kick off tab to assist removal
  • Antistatic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
40
Reusable
Unisex
NORA
Trainers
S5
ESD Safety Shoe
PVC
Rubber
EN ISO 20345:2011
Standard Width
Yes
No
SRC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.