Learn More
Tocris Bioscience™ Noradrenaline bitartrate
Endogenous adrenergic hormone and neurotransmitter
Brand:  Tocris Bioscience™ 5169/50
77.75 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18707486
  • 102.00 € / 50mg
Explore more special offers
Description
Description
Noradrenaline bitartrate is an endogenous adrenergic hormone and neurotransmitter. A classical stress hormone; underlies the fight or flight response, along with adrenaline. Increases heart rate. Vasoconstrictor.
Specifications
Specifications
>99%
50 mg
319.26
Norepinephrine
Noradrenaline bitartrate
Noradrenaline bitartrate