Learn More
Norbornene, 99%, Thermo Scientific Chemicals
Description
Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs' catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs′ catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.
Solubility
Soluble in ethanol, ether and most organic solvents. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 44°C to 47°C |
Density | 0.955 |
Boiling Point | 95°C to 96°C |
Flash Point | −15°C (5°F) |
Odor | Pungent |
Quantity | 500 g |
UN Number | UN1325 |
Beilstein | 1098544 |
Solubility Information | Soluble in ethanol,ether and most organic solvents. Insoluble in water. |
Formula Weight | 94.16 |
Show More |
RUO – Research Use Only