missing translation for 'onlineSavingsMsg'
Learn More

Norell™ Standard Series™ 10mm Precision NMR Sampling Tubes

Excellent for monitoring reactions, scouting for new compounds or doing routine NMR. Norell™ Standard Series™ 10mm Precision NMR Sampling Tubes have been developed for quality and precision and are the top choice for today's scientist.

79.05 € - 151.00 €

Specifications

Material Borosilicate Glass
Product Type Ultra Precision NMR Sample Tube
For Use With (Application) Routine NMR
Includes Cap
Quantity 5 Pack
View More Specs

Products 3
Product Code Brand Length (Metric) Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Outer Price Quantity & Availability  
12852296
View Documents
Norell™
508-UP-7
178 mm 4.97 mm
79.05 €
Pack of 5
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
12862296
View Documents
Norell™
508-UP-8
203 mm 4.97 mm
102.00 €
Pack of 5
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
12822336
View Documents
Norell™
1005-P-7
178 mm 10 mm
151.00 €
Pack of 5
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
Description

Description

  • For auto-sampling systems and high-throughput NMR
  • Polished ends to make tubes less susceptible to breaking
  • Colored caps included
  • Virtually all bottom and wall variation has been eliminated, greatly improving shimmering times and sample reproducibility
Specifications

Specifications

Borosilicate Glass
Routine NMR
5 Pack
Ultra Precision NMR Sample Tube
Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.