missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Normal Goat Serum
Normal Goat Serum
Brand: Gibco™ PCN5000
106.40 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Normal goat serum. Whole serum is obtained from non-immunized (preimmune) healthy goats. The serum is clarified through a 0.8 micron low protein binding filter.
Specifications
Store in freezer (-5 to -30°C). | |
Liquid | |
5 mL | |
Dry Ice |
Goat/Caprine | |
Goat Serum | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Normal Goat Serum
Version: 1.00
File size: 87.5 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2448562 | Certificate of Analysis | 29/01/2025 | PCN5000 |
2507428 | Certificate of Analysis | 29/01/2025 | PCN5000 |
21118007 | Certificate of Analysis | 28/01/2025 | PCN5000 |
231016015 | Certificate of Analysis | 28/01/2025 | PCN5000 |
240319011 | Certificate of Analysis | 27/01/2025 | PCN5000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Normal Goat Serum > 5mL
Analyte Specific Reagent. Analytical and performance characteristics are not established.
Spot an opportunity for improvement?Share a Content Correction