missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ North2South™ Chemiluminescent Substrate for HRP

Acquire consistent, highly sensitive Southern blot results with a novel ECL HRP substrate and optimized hybridization and blocking buffers.

Brand:  Thermo Scientific™ 17295

463.60 EUR valid until 2024-12-31
Use promo code "21954" to get your promotional price.


Product Code. 11876704

  • 488.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ North2South Chemiluminescent Detection Kit combines a novel ECL substrate for horseradish peroxidase (HRP) and optimized hybridization and blocking buffers to ensure consistent, high-sensitivity Southern blot results.

The Thermo Scientific North2South Hybridization and Detection Kit combines a novel enhanced luminol substrate for horseradish peroxidase (HRP) with optimized pre-made hybridization and blocking buffers to ensure consistent results and high sensitivity. Any nucleic acid blot can be hybridized with the appropriate biotinylated (biotin-labeled) oligonucleotide probe. After a brief membrane blocking step, the biotin label is detected with streptavidin-HRP and a 5-minute incubation with the chemiluminescent substrate. Results are easily recorded by brief exposure to X-ray film (1-15 minutes) or CCD camera.

Highlights:

  • Sensitivity equal to or greater than 32P
  • Amenable to many formats
  • High level of light output and consistent low background allows for very short exposure times using cooled CCD camera systems or film
  • Ready-to-use buffers and short process time makes switching from radioactive detection to chemiluminescent detection an easy transition, even for the novice user
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient For: 1000 cm2 of membrane (10 mini-gel blots)
• Luminol/Enhancer Solution, 50 mL
• Stable Peroxide Solution, 50 mL

Store at 4°C.
North2South™
100 mL Kit
Chromogenic
Substrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers