Learn More
Merck Millipore Novagen™ KOD Hot Start DNA Polymerase
For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications
729.00 €
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12838435
View Documents
Merck Millipore
71086-3
729.00 €
Each
View alternative products
Save Up to 
»
Description
Description
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA templates up to 21 kbp
  • Successfully amplifies GC-rich sequences
  • Eliminates mispriming and primer-dimer formation
  • Convenient room-temperature setup compatible with automation
  • Optimal KOD Hot Start Buffer for PCR performance over wide range of targets