Learn More
Novex™ Novex™ Zymogram Plus (Gelatin) Protein Gels, 10%, 1.0 mm
Novex 10% Zymogram Plus (Gelatin) gels are useful for the detection and characterization of proteases that use gelatin as a substrate. The proteases are run under denaturing conditions and visualized as clear bands against a dark background using a simple renaturing, developing, and staining protocol.
Brand: Novex™ ZY00105BOX
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
Novex 10% Zymogram Plus (Gelatin) gels are useful for the detection and characterization of proteases that use gelatin as a substrate. The proteases are run under denaturing conditions and visualized as clear bands against a dark background using a simple renaturing, developing, and staining protocol. Zymogram gels are commonly used to detect matrix metalloproteinases. Novex 10% Zymogram Plus gels are highly sensitive, detecting as little as 5 x 10-6 units of collagenase.
Formulation
Novex Zymogram Plus gels are Tris-glycine gels with 0.1% gelatin incorporated into the gel. They are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, highly purified water, and gelatin as the substrate. All have a 4% stacking gel.
Choose the right gel format for your experiments
Novex 10% Zymogram Plus gels come in a fixed concentration of 10% gelatin and three different well formats, including 10-, 12- and 15-well.
Recommended buffers
Use Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC2675) with these gels. Novex Zymogram Renaturing Buffer (LC2670) and Novex Zymogram Developing Buffer (LC2671) are also recommended.
Recommended Storage
Store in refrigerator (2-8°C). Do not freeze.
Specifications
Mini Gel Tank, XCell SureLock™ Mini | |
Novex | |
Zymogram Gel | |
15-well | |
Up to 15 μL | |
Wet Ice | |
1.0 mm |
Mini | |
0.1 | |
Molecular Weight | |
10 Gels/Box | |
30 to 150 kDa (Tris-Glycine Buffer) | |
Store at 2°C to 8°C. Do not freeze. | |
8 cm |